Orders LinkContactUsLinkComponent
Precision Screwdriver Set, Electron Microscopy Sciences
Precision Screwdriver Set, Electron Microscopy Sciences
Replacement Blade, 1.2 mm
Catalog # 100497-590
Supplier #:  62078-12
Precision Screwdriver Set, Electron Microscopy Sciences
Catalog # 100497-590
Precision Screwdriver Set, Electron Microscopy Sciences
Precision Screwdriver Set, Electron Microscopy Sciences
Replacement Blade, 1.2 mm
Price
$6.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The hexagonal swivel heads prevent rolling and knurled handles for a firm grip. Color identifier bands allow for easy selection and set screws hold the blades securely. Set contains 19 extra blades and 2 extra set screws.
Delivery information: The set comes with a handsome revolving stand.
Specifications
  • Description:
    Replacement Blade
  • Size:
    1.2 mm