Orders LinkContactUsLinkComponent
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Glennel®, Diamond Suspension, Electron Microscopy Sciences
0.5 µm
  100497-346
Supplier #:  50372-30
Glennel®, Diamond Suspension, Electron Microscopy Sciences
  100497-346
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Glennel®, Diamond Suspension, Electron Microscopy Sciences
0.5 µm
$108.91
Each
Glennel® Diamond Suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring.
  • A versatile, water based suspension designed for most production applications
  • Excellent lubrication
  • Uniform fast lapping action
  • Maximum cutting action
  • Compatible with all common extenders
The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators.
Packaging: All Diamond Suspensions are packed in 400 g bottles and have Medium Concentration prepared with Premium Polycrystalline Diamond in either Propylene Glycol (Oil based) or Water based formulas.
  • Size:
    0.5 µm