Partitioned Plastic Boxes, Electron Microscopy Sciences
Product Details & Documents
Organize singular or groups of items in individual compartments.
- Extra-thick wall construction permits containment of heavy objects, protects delicate items
Stored items stay separately and are readily visible at all times.
Specifications
- Description:12 Compartments
- Dimensions:7×3-3/4×1-1/4" (Overall); 1-11/16×1-1/16×1-1/8" (Compartments)