Orders LinkContactUsLinkComponent
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Block of 40 Standards X-Ray
Catalog # 100495-022
Supplier #:  80070-40
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Catalog # 100495-022
CAS Number:  
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Block of 40 Standards X-Ray
CAS Number:  
Price
$10,437.41
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
  • All standards are polished to a 0.25 micron diamond finish and carbon coated
  • They are supplied with fully authenticated certificates of analyses and a location map for standard identification
Pure metals and compounds mounted in brass blocks. Carousel configuration is supplied at an extra cost of (52.00) to the above prices. Please note that for this configuration there is a maximum of 25 standards permitted.
A Faraday Cup, for accurate specimen current measurements, is available as an optional extra on all mounts. The standards are available in different sizes of brass holders, including those of 25 or 32 mm diameter, containing up to 60 standards of your choice. Carousels, 13 mm diameter blocks (to fit pin stubs), and single standards of 5 mm, 3 mm, 2 mm diameter, are also available. Specials can be "tailor-made" to suit individual needs and specific stage configurations.
Specifications
Specifications
  • Description:
    Block of 40 Standards X-Ray