Orders LinkContactUsLinkComponent
Carolina ™ Blue Box, Electron Microscopy Sciences
Carolina ™ Blue Box, Electron Microscopy Sciences
Carolina Blue Box, 25 Slide
Catalog # 100493-364
Supplier #:  71483-01
Carolina ™ Blue Box, Electron Microscopy Sciences
Catalog # 100493-364
Carolina ™ Blue Box, Electron Microscopy Sciences
Carolina ™ Blue Box, Electron Microscopy Sciences
Carolina Blue Box, 25 Slide
Price
$11.19
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Molded from blue high-impact styrene, the Carolina Slide Box is manufactured to exacting specifications.
Compared to conventional wooden boxes, the Carolina Blue Box is smaller in size, lighter in weight, and held to closer dimensional tolerances. The 75 x 25 mm (3 x 1") slides fit the slots and will not slip out once in place. A shoulder on each box excludes dust and identities the top of the box. The boxes are available in two sizes: 3 1/4 x 4 1/4 x 1 3/16" (for 12 slides).
Specifications
  • Capacity:
    25 Slide
  • Description:
    Carolina Blue Box