Orders LinkContactUsLinkComponent
Low Intensity UV Lamp, Electron Microscopy Sciences
Low Intensity UV Lamp, Electron Microscopy Sciences
Low-Intensity Bulb
Catalog # 100491-964
Supplier #:  72425
Low Intensity UV Lamp, Electron Microscopy Sciences
Catalog # 100491-964
Low Intensity UV Lamp, Electron Microscopy Sciences
Low Intensity UV Lamp, Electron Microscopy Sciences
Low-Intensity Bulb
Price
$60.36
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements.
Constructed from Cyolac® plastic, with a longwave emission of 365 nm and an intensity at 6" (15.2 cm) of 750 µm/cm². The bulb has a rated life of 6000 hours. It can be used as a hand-held light or it may br mounted on a optional stand. Powered by 110 V/60 Hz or 220 V/50 Hz.
Specifications
  • Description:
    Low-Intensity Bulb