Numbered Grid Storage Box, 100 Capacity, Electron Microscopy Sciences
Product Details & Documents
This is the standard 100 capacity grid storage box with a unique number printed on the face and on one end.
- Eliminates the placement of the specimen grid in the wrong box
- Easy retrieval of grid box from storage
- Complete with grid recording card
Specifications
- Description:Numbered Grid Storage Box
- Size:1/Pack