Orders LinkContactUsLinkComponent
Cutting Mats, Electron Microscopy Sciences
Cutting Mats, Electron Microscopy Sciences
Cutting Mat 6 x 8"
Catalog # 100491-558
Supplier #:  69037
Cutting Mats, Electron Microscopy Sciences
Catalog # 100491-558
Cutting Mats, Electron Microscopy Sciences
Cutting Mats, Electron Microscopy Sciences
Cutting Mat 6 x 8"
Price
$48.87
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cutting mats are inert, self-healing and can be sterilized with ethanol. Available sizes 6 x 8" and 21/2 x 3".
Specifications
  • Description:
    Cutting Mat 6 x 8"