Orders LinkContactUsLinkComponent
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
30 ml (1 oz.), 25 mm
Catalog # 100490-544
Supplier #:  64264-03
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Catalog # 100490-544
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
30 ml (1 oz.), 25 mm
Price
$359.85
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These lightweight transparent PVC sample bottles come with snap-on tight-fitting caps and they offer excellent clarity, durability, and good chemical resistance. Maximum temperature –20 to 80 °C (–4 to 176 °F).
Note: The following sizes are also available: 30, 40, 48, 60, 90, 120,and 180 ml.
Specifications
  • Capacity:
    30 ml (1 oz.)
  • Height:
    25 mm