Orders LinkContactUsLinkComponent
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
5 Dram (18.5 ml), 40 mm, 22 mm
Catalog # 100490-530
Supplier #:  64248-15
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Catalog # 100490-530
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
5 Dram (18.5 ml), 40 mm, 22 mm
Price
$23.77
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
Specifications
  • Ø int.:
    22 mm
  • Capacity:
    5 Dram (18.5 ml)
  • Height:
    40 mm