Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tissue Processing Capsules, Electron Microscopy Sciences

Tissue Processing Capsules, Electron Microscopy Sciences

Tissue Processing Capsule for Biopsy Tissue, White, 29 mm, 6 mm
Catalog # 100490-176
Supplier #:  62359-W
Tissue Processing Capsules, Electron Microscopy Sciences
Catalog # 100490-176
Tissue Processing Capsules, Electron Microscopy Sciences
Tissue Processing Capsules, Electron Microscopy Sciences

Tissue Processing Capsule for Biopsy Tissue, White, 29 mm, 6 mm

Price
$229.04
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These one piece plastic capsules are hinged and perfectly aligned, offering convenience and ease of use

Both the top and the bottom of the capsule have an open mesh offering maximum fluid flow during processing. A frosted marking surface assures permanent identification. These capsules are commonly used with the Embedding Ring System. They are available in two sizes and five colors.

Specifications
  • Depth:
    6 mm
  • Ø:
    29 mm
  • Color:
    White
  • Description:
    Tissue Processing Capsule for Biopsy Tissue

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.