Orders LinkContactUsLinkComponent
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Breaker, 10 - 20 ml ampoules
  100490-064
 :  60605-20
Ampoule Cracker, Electron Microscopy Sciences
  100490-064
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Breaker, 10 - 20 ml ampoules
$190.61
Pack of 500
The Ampoule Cracker™ is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules.
Simply slip the Ampoule Cracker™ over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
  • Description:
    Ampoule Breaker
  • For:
    10 - 20 ml ampoules