Orders LinkContactUsLinkComponent
Cryo Markers, Electron Microscopy Sciences
Cryo markers
  100489-748
Cryo Markers, Electron Microscopy Sciences
  100489-748
Cryo Markers, Electron Microscopy Sciences
Cryo Markers, Electron Microscopy Sciences
Cryo markers
$41.45
Pack of 4
Used for permanently marking cloth, cardboard, and plastic.
It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).
  • Description:
    Cryo markers