Orders LinkContactUsLinkComponent
View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
Refill View Pack
Catalog # 100489-338
Supplier #:  71544-50
View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
Catalog # 100489-338
View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
Refill View Pack
Price
$86.60
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For the orderly visual display and safe transport of microscope slides.
This loose-leaf binder has ten vinyl View-Pack pages (hold 160 slides). Each page holds 16 slides 75 x 25 mm (3 x 1") in individual pockets with a white back and clear front. A flap in the center keeps slides from falling out. Simply fold the page at the center crease to retrieve slides. The 22 x 27 cm (8 1/2 x 10 1/2") View-Pack pages comes with a standard 23 x 30 cm (9 x 12") 3-ring binder.
Specifications
  • Description:
    Refill View Pack