Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

LEO SEM Mount, Electron Microscopy Sciences

LEO Microscope Mounts Slotted Head, 50
  100488-660
LEO SEM Mount, Electron Microscopy Sciences
  100488-660
LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences

LEO Microscope Mounts Slotted Head, 50

 
$53.18
   
Pack of 50
 
   
 

These mounts with tapered end pin

  • Slotted head 12.7 mm (1/2") diameter
  • Pin 3.1 x 5 mm (1/8 x 1/4")
 
  • Description:
    LEO Microscope Mounts Slotted Head
  • Size:
    50

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.