Orders LinkContactUsLinkComponent
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Polystyrene slides
Catalog # 100488-460
Supplier #:  71889
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Catalog # 100488-460
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Polystyrene slides
Price
$652.50
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Note: Long exposure to solvents will cause the slide to warp
Permanox™ is a strong, biologically inert material resistant to non-chlorinated hydrocarbons, acetone, and other EM reagents. Permanox™slides allow you to do cell fixation and staining on the slide; they can be cut with scissors. Permanox™ has not been shown to be autofluorescent. All of the microscope slides are 1 x 3".
Note: Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Specifications
  • Description:
    Polystyrene slides