Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Anionic Gold Tracer, Electron Microscopy Sciences

Anionic Gold Tracer, Electron Microscopy Sciences

EM Anionic Gold Tracer, 5 ml, 25 nm
Catalog # 100487-900
Supplier #:  25473
Anionic Gold Tracer, Electron Microscopy Sciences
Catalog # 100487-900
CAS Number:  
Anionic Gold Tracer, Electron Microscopy Sciences
Anionic Gold Tracer, Electron Microscopy Sciences

EM Anionic Gold Tracer, 5 ml, 25 nm

CAS Number:  
Price
$289.99
Log in for availability
Each (5ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Anionic tracers are used to visualize positive charged moieties in specimens

  • Particle sizes of 0 8, 6, 10, 15 and 25 nm
  • Guaranteed shelf life of 18 months
  • Supplied in PBS with 15 mM NaN3

Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).

Specifications

Specifications

  • Description:
    EM Anionic Gold Tracer
  • Size:
    25 nm
  • Volume:
    5 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.