$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
MP-285 MECHANICAL ONLY
100358-428
:
Sutter Instruments Company
MP-285 MECHANICAL ONLY
100358-428
:
Sutter Instruments Company
:
MP-285/M