Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MP285 SYSTEM,TABLE MT
Catalog # 100358-256
Supplier:
Sutter Instruments Company
Expand image
MP285 SYSTEM,TABLE MT
Catalog # 100358-256
Supplier:
Sutter Instruments Company
Supplier Number:
MP-285/T