AMV Reverse Transcriptase, New England Biolabs
AMV Reverse Transcriptase, 1,000 units
101417-974
: M0277L
Avian Myeloblastosis Virus (AMV) Reverse Transcriptase is an RNA-directed DNA polymerase
This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template.
- Description:AMV Reverse Transcriptase
- Size:1,000 units