Orders LinkContactUsLinkComponent
General Purpose Transfer Pipette, Electron Microscopy Sciences
General Purpose Transfer Pipette, Electron Microscopy Sciences
4.0 ml, 6.6×15.4 cm, Sterile, 1.9 ml
Catalog # 101413-278
Supplier #:  70960-5S
General Purpose Transfer Pipette, Electron Microscopy Sciences
Catalog # 101413-278
General Purpose Transfer Pipette, Electron Microscopy Sciences
General Purpose Transfer Pipette, Electron Microscopy Sciences
4.0 ml, 6.6×15.4 cm, Sterile, 1.9 ml
Price
$128.36
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These pipettes are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipettes are bulk packed and the sterile are individually packed.
Specifications
  • Ø×L:
    6.6×15.4 cm
  • Sterility:
    Sterile
  • Volume:
    4.0 ml
  • Bulb draw:
    1.9 ml