Orders LinkContactUsLinkComponent
Pasteboard Sliding-Boxes, Electron Microscopy Sciences
Pasteboard Sliding-Boxes, Electron Microscopy Sciences
Paste Board Box, 82×57×34 mm (E×terior), 80×53×31 mm (Interior)
Catalog # 101412-974
Supplier #:  64326-11
Pasteboard Sliding-Boxes, Electron Microscopy Sciences
Catalog # 101412-974
Pasteboard Sliding-Boxes, Electron Microscopy Sciences
Pasteboard Sliding-Boxes, Electron Microscopy Sciences
Paste Board Box, 82×57×34 mm (E×terior), 80×53×31 mm (Interior)
Price
$105.23
Log in for availability
Pack of 72
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These sliding-boxes are ideal for the storage of specimen blocks and small items.
  • Constructed from white pasteboard
  • Match box style and rectangular in shape
  • Available in different sizes to meet your needs
Specifications
  • Description:
    Paste Board Box
  • Dimensions:
    82×57×34 mm (E×terior), 80×53×31 mm (Interior)