Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FORM - STRAIGHT BILL OF LADING
Catalog # 101376-190
Supplier:
Label Master
Expand image
FORM - STRAIGHT BILL OF LADING
Catalog # 101376-190
Supplier:
Label Master
Supplier Number:
F360-4