Orders LinkContactUsLinkComponent
MeshWell Plates, Electron Microscopy Sciences
MeshWell Plates, Electron Microscopy Sciences
Plate 24 well
Catalog # 100502-548
Supplier #:  64700-24
MeshWell Plates, Electron Microscopy Sciences
Catalog # 100502-548
MeshWell Plates, Electron Microscopy Sciences
MeshWell Plates, Electron Microscopy Sciences
Plate 24 well
Price
$350.32
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500 µm opening.
  • Available with 6, 12, and 24 well configurations
  • Also able to process tissue slices with Reagent Trays
Fits into the 6, 12 and 24 Costar/Corning Cell Culture Clusters for processing tissue slices.
Ordering information: The MeshWell Plate set includes one MeshWell and one Cluster plate.
Specifications
  • Description:
    Plate 24 well