Orders LinkContactUsLinkComponent
Broken Crosslines, Electron Microscopy Sciences
Broken Crosslines, Electron Microscopy Sciences
NE56 Broken Crosslines Graticule, 19 mm
Catalog # 100499-028
Supplier #:  67985-19
Broken Crosslines, Electron Microscopy Sciences
Catalog # 100499-028
Broken Crosslines, Electron Microscopy Sciences
Broken Crosslines, Electron Microscopy Sciences
NE56 Broken Crosslines Graticule, 19 mm
Price
$199.30
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Broken lines enable fine detail to be seen at the breaks. A thin boundary would be lost behind a continuous line.
Specifications
  • Description:
    NE56 Broken Crosslines Graticule
  • Size:
    19 mm