Netwell™ Tissue Processing Well, Electron Microscopy Sciences
Product Details & Documents
Home made devices are no longer necessary
These tissue processing wells are ready to fill all of your needs with the added convenience of being able to process up to 12 tissue specimens at one time (with one unit) . Tissue processing well is a modular and disposable system which can be used for the following procedures:
The handling and processing of tissue slices.
In immunocytochemical staining.
In 3-dimensional tissue culture.
In explant tissue culture.
Home made devices are no longer necessary. These tissue processing wells are ready to fill all of your needs with the added convenience of being able to process up to 12 tissue specimens at one time (with one unit) .
The Netwell™ is a polystyrene insert fitted with a polyester mesh bottom for use in 6 and 12-well plates. Available in three mesh sizes and two diameters; the Netwells provide ultimate flexibility in retaining fragile tissues during processing and staining.
Netwell™ Carrier Kit is a set of carrier plates and handles for moving the Netwells in and out of the 6 or 12-well plates. The handle can be attached or detached to the plate at any time.
Netwell™ Reagent Tray is a thermo-formed polystyrene. Holds 150 m of reagent solution. Netwells are trans-ferred to the reagent tray by the Carrier kit, where fixing and staining is performed. Reagent trays are available in white for colorimetric reaction contrast, and in black for better visibility of tissue sections.
Applications: The polyester mesh acts as a tissue carrier and strainer for immunocytochemical staining and biological and histological fixation. It avoids the risk of losing valuable samples associated with paint-brush transfer. For 3-D tissue culture, the sterile Netwell provides efficient media permeation and support to cultures grown in gels. As well, Sterile Netwells allow for easy processing of tissue explants and microcarrier-grown cells. For your convenience, the Netwell™ Sets are available ready to use.
The set consists of:
6 Netwells of 24mm diameter or 12 Netwells of 15mm diameter.
Two Reagent Trays, one black and one white.
One Carrier Kit: one carrier plate and one handle. The Netwell™ is a polystyrene insert fitted with a polyester mesh bottom for use in 6 and 12-well plates. Available in three mesh sizes and two diameters; the Netwells provide ultimate flexibility in retaining fragile tissues during processing and staining.
Netwell™ Carrier Kit is a set of carrier plates and handles for moving the Netwells in and out of the 6 or 12-well plates. The handle can be attached or detached to the plate at any time.
Netwell™ Reagent Tray is a thermo-formed polystyrene. Holds 150 m of reagent solution. Netwells are trans-ferred to the reagent tray by the Carrier kit, where fixing and staining is performed. Reagent trays are available in white for colorimetric reaction contrast, and in black for better visibility of tissue sections.
Applications: The polyester mesh acts as a tissue carrier and strainer for immunocytochemical staining and biological and histological fixation. It avoids the risk of losing valuable samples associated with paint-brush transfer. For 3-D tissue culture, the sterile Netwell provides efficient media permeation and support to cultures grown in gels. As well, Sterile Netwells allow for easy processing of tissue explants and microcarrier-grown cells. For your convenience, the Netwell™ Sets are available ready to use.
The set consists of:
6 Netwells of 24mm diameter or 12 Netwells of 15mm diameter.
Two Reagent Trays, one black and one white.
One Carrier Kit: one carrier plate and one handle.
Specifications
- Description:Set
- Size:15 mm
- Sterile:Sterile
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



