Orders LinkContactUsLinkComponent
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 2 x 70 mm, 26 Gauge
  100498-104
 :  62437-02
Inoculating Loops, Electron Microscopy Sciences
  100498-104
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 2 x 70 mm, 26 Gauge
$277.34
Each
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
  • Description:
    Inoculating Loops
  • Size:
    2 x 70 mm
  • Gauge:
    26 Gauge