Diamond Points, Electron Microscopy Sciences
Product Details & Documents
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Specifications
- Ø:3.8 mm
- Description:Diamond Points, Square Grid
- Shape:Cone