Precision Screwdriver Set, Electron Microscopy Sciences
Product Details & Documents
The hexagonal swivel heads prevent rolling and knurled handles for a firm grip. Color identifier bands allow for easy selection and set screws hold the blades securely. Set contains 19 extra blades and 2 extra set screws.
Delivery information: The set comes with a handsome revolving stand.
Specifications
- Description:Replacement Blade
- Size:3.0 mm