Flat Mounting Blocks , Electron Microscopy Sciences
Product Details & Documents
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.
Specifications
- Ø:1.25"
- Description:Flat Mounting Blocks (Stainless Steel)