Orders LinkContactUsLinkComponent
X-Checker™, Electron Microscopy Sciences
X-Checker™, Electron Microscopy Sciences
X-Checker, Extra
Catalog # 100495-000
Supplier #:  80058-EX
X-Checker™, Electron Microscopy Sciences
Catalog # 100495-000
CAS Number:  
X-Checker™, Electron Microscopy Sciences
X-Checker™, Electron Microscopy Sciences
X-Checker, Extra
CAS Number:  
Price
$1,687.86
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The X-Checker™ was the first and remains the only complete calibration aid for SEM/EDS Systems.
Each X-Checker comes with manganese to measure full width at half max detector resolution, aluminum and copper to perform spectral calibration, and carbon to monitor calibration at the low end of the spectra for thin window detectors. You also get two grid sizes for checking the accuracy of your image analysis software and an easy test for monitoring the amount of vacuum pump oil contamination on your detector window.
The X-Checker™ BN comes with boron nitride for those who need a more sensitive monitor of low end performance on thin window and windowless detectors.
The X-Checker™ Extra is the ultimate performance monitor for the latest state of the art X-ray detectors. In addition to the standard features and boron nitride, there is a fluorine source to test resolution at the fluorine K-alpha peak (industry standard for measuring low end resolution). As well it comes with a beryllium grid for the ultimate test of detector performance.
Specifications
Specifications
  • Description:
    X-Checker, Extra