Orders LinkContactUsLinkComponent
Burnishers, Electron Microscopy Sciences
Burnishers, Electron Microscopy Sciences
Standard Tip Burnisher
Catalog # 100494-388
Supplier #:  77615-01
Burnishers, Electron Microscopy Sciences
Catalog # 100494-388
Burnishers, Electron Microscopy Sciences
Burnishers, Electron Microscopy Sciences
Standard Tip Burnisher
Price
$24.65
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These burnishers are especially designed for dry transfer products. They are pressure adjustable allowing you to set and maintain the proper pressure for any burnishing style.
For maximum adhesion, place the backing sheet over the newly transferred letters and reburnish with the "bone end." The rolling tip burnisher is ideal for large transfers, the standard tip is ideal for standard work, and the fine tip for detail work.
Specifications
  • Description:
    Standard Tip Burnisher