Orders LinkContactUsLinkComponent
High Opacity Paint Markers, Electron Microscopy Sciences
High Opacity Paint Markers, Electron Microscopy Sciences
Large Marker, White
Catalog # 100493-640
Supplier #:  72168-W2
High Opacity Paint Markers, Electron Microscopy Sciences
Catalog # 100493-640
High Opacity Paint Markers, Electron Microscopy Sciences
High Opacity Paint Markers, Electron Microscopy Sciences
Large Marker, White
Price
$10.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface
  • Weatherproof
  • Resistant to alcohol and acetone
  • Contain xylene
Markers are available in black, red and white with bullet tips. Except model 130, it writes like a pen with white ink for marking on dark surfaces, including carbon tabs, which is use in SEM specimen mounting purpose.
Specifications
  • Description:
    Large Marker, White