Graduated Pitchers, Electron Microscopy Sciences
Product Details & Documents
Gracefully proportioned, molded pitchers with large comfortable handles and well designed pouring spouts.
- Metric graduations, and crystal clear to the eye (polymethylpentene)
- Resists heat to 175°C (374°F)
- Shatter-proof
- Excellent chemical resistance
Ideal for preparation of photographic chemicals.
Specifications
- Capacity:1000 ml