Orders LinkContactUsLinkComponent
Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Grid Storage Box, Clear Cover, 50 Grids
Catalog # 100491-744
Supplier #:  71150
Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Catalog # 100491-744
Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Grid Storage Box, Clear Cover, 50 Grids
Price
$12.30
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Documents
Specifications
  • Capacity:
    50 Grids
  • Description:
    Grid Storage Box, Clear Cover