Orders LinkContactUsLinkComponent
Dust-Off Plus Model (DPN), Electron Microscopy Sciences
Dust-Off DPN Kit
  100491-658
Dust-Off Plus Model (DPN), Electron Microscopy Sciences
  100491-658
Dust-Off Plus Model (DPN), Electron Microscopy Sciences
Dust-Off Plus Model (DPN), Electron Microscopy Sciences
Dust-Off DPN Kit
$110.27
Each
Contains formula N (tetrafluoroethane).
  • 100% safe for the ozone
  • Contains no flammable elements
It is provided with a patented 360° vector valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas.
Delivery information: The kit consists of a 8 oz. can and a vector valve nozzle.
  • Description:
    Dust-Off DPN Kit