Tungsten Wire Filaments, Electron Microscopy Sciences
Product Details & Documents
V-shaped filaments are high purity, vacuum grade tungsten wire 0.02" (0.50 mm) diameter, V-shaped filaments ½" (12.7mm) deep, with a bottom angle of 45°, either single wire or triple strand.
Coiled filaments are single or 3-strand coiled tungsten filaments, 0.5 mm (0.002") diameter wires.
Specifications
- Ø:0.5 mm (0.02")
- Description:Triple tungsten V-shaped filaments, 5/8" (16 mm) v-opening
- Length:3"