Orders LinkContactUsLinkComponent
Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences
0.2% Formaldehyde/0.1% Chromic Acid in 0.1 M Sodium Cacodylate Buffer (pH 7.4)
Catalog # 100503-950
Supplier #:  15953
Fixatives, Prepared, Electron Microscopy Sciences
Catalog # 100503-950
CAS Number:  
Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences
0.2% Formaldehyde/0.1% Chromic Acid in 0.1 M Sodium Cacodylate Buffer (pH 7.4)
CAS Number:  
Price
$75.23
Log in for availability
Each (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.
 : Certificates of analysis are available.
Specifications
Specifications
  • Description:
    0.2% Formaldehyde/0.1% Chromic Acid in 0.1 M Sodium Cacodylate Buffer (pH 7.4)