Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences

RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences

Red-Z Fluid Control Solidifier, 11 oz.
Catalog # 100503-194
Minority or Woman-Owned Business Enterprise
Supplier #:  24145-10
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
Catalog # 100503-194
Minority or Woman-Owned Business Enterprise
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences

Red-Z Fluid Control Solidifier, 11 oz.

Price
$48.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

RED-Z Solidifier will solidify, decontaminate and deodorize any water base fluid at a ratio of ¾ oz to 1 L

It contains chlorine (NaDCC) an EPA registered disinfectant - almost twice the amount recommended by the CDC. Solidifier could also be used for blood and other body fluids. RED-Z available in a pointed top bottle that fits easily in porthole containers and in a flip-top shaker top bottle for large spills.

Specifications
  • Capacity:
    11 oz.
  • Description:
    Red-Z Fluid Control Solidifier
  • Cat. No.:
    100503-194

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.