Orders LinkContactUsLinkComponent
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Stub Assembly
Catalog # 100503-044
Supplier #:  50116-65
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Catalog # 100503-044
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Stub Assembly
Price
$104.34
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A Tripod Polisher® was designed to prepare accurately micro sizes of TEM and SEM samples.
  • Limited ion milling
  • No strong chemicals are used to prepare the specimen
  • Reduce preferential thinning, radiation damage, and heating of the sample
Used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals and, geological specimens. The same sample can be studied with both SEM and TEM and hence reduces sample preparation.
Specifications
  • Description:
    Stub Assembly