Orders LinkContactUsLinkComponent
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
25 Capacity
Catalog # 100501-096
Supplier #:  71460
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Catalog # 100501-096
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
25 Capacity
Price
$29.96
Log in for availability
Bag of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from durable polystyrene (with heavy walls).
Same characteristics as the Secured Hinge Microscope Slide Boxes, but with a slip-on cover. Holds 25 1x3" slides. Measures H x W x L: 1/4 x 3 1/2 x 5 1/2".
Specifications
  • Description:
    25 Capacity