Orders LinkContactUsLinkComponent
Process Embedding Cassettes, Electron Microscopy Sciences
Process Embedding Cassettes, Electron Microscopy Sciences
Process/Embedding Cassettes w/o Lids, Tan
Catalog # 100500-550
Supplier #:  70025-T
Process Embedding Cassettes, Electron Microscopy Sciences
Catalog # 100500-550
Process Embedding Cassettes, Electron Microscopy Sciences
Process Embedding Cassettes, Electron Microscopy Sciences
Process/Embedding Cassettes w/o Lids, Tan
Price
$95.34
Log in for availability
Case of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from Acetal copolymer – Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are molded from a high-density polymer that is totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Only Clear cassettes are made of polypropylene. All other cassettes are made of Acetal Polymer. Each case contains 3 dispenser boxes of 500 cassettes. Stainless Steel Lids order separately.
Specifications
  • Color:
    Tan
  • Description:
    Process/Embedding Cassettes w/o Lids