Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences

Gold Seal Plain Microscope Slides, Electron Microscopy Sciences

Plain slides, 1 g, 1.0 mm, 75×25 mm (3×1")
Catalog # 100498-534
Supplier #:  63710-25
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Catalog # 100498-534
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences

Plain slides, 1 g, 1.0 mm, 75×25 mm (3×1")

Price
$1,346.46
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.

To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.

Specifications
  • Capacity:
    1 g
  • Description:
    Plain slides
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.