Orders LinkContactUsLinkComponent
Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Loop and Needle Holder, 203 mm (8")
Catalog # 100498-116
Supplier #:  62440-08
Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Catalog # 100498-116
Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Loop and Needle Holder, 203 mm (8")
Price
$247.82
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The innoculating loops are made from durable, heat resistant brass with a fiber glass insulating sleeve over the gripping area. The threaded collet type collar holds all wire gauge.
Specifications
  • Description:
    Loop and Needle Holder
  • Length:
    203 mm (8")