Orders LinkContactUsLinkComponent
Diamond Points, Electron Microscopy Sciences
Diamond Points, Inverted, Cone
  100497-974
 :  62155-23
Diamond Points, Electron Microscopy Sciences
  100497-974
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Inverted, Cone
$37.91
Each
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
  • Description:
    Diamond Points, Inverted
  • Shape:
    Cone