Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Product Details & Documents
A versatile hang-up flexible shaft machine.
Accessories information: Mounted accessories are on 2.35 mm (3/32") arbors.
Ordering information: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
Specifications
- Description:1/10 Hp Flex Shaft Motor
- Voltage:220 V
Accessories
Product Information
- Catalog #AvailabilityPrice
- Standard Foot Speed Control
Product Information
- Catalog #AvailabilityPrice
- Standard Foot Speed Control
Product Information
- Catalog #AvailabilityPrice
- Probe Handle
Product Information
- Catalog #AvailabilityPrice
- Replacement Flex Shaft
Product Information
- Catalog #AvailabilityPrice
- Replacement Sheath
Product Information
- Catalog #AvailabilityPrice
- 13-Piece Assortment Set
Product Information
- Catalog #AvailabilityPrice
- 26-Piece Assortment Set
Product Information
- Catalog #AvailabilityPrice
- Flex Shaft Stand