Orders LinkContactUsLinkComponent
PTFE Beakers, Electron Microscopy Sciences
PTFE Beakers, Electron Microscopy Sciences
1 ml, PTFE Beaker Cover, White
Catalog # 100497-462
Supplier #:  60936-C
PTFE Beakers, Electron Microscopy Sciences
Catalog # 100497-462
PTFE Beakers, Electron Microscopy Sciences
PTFE Beakers, Electron Microscopy Sciences
1 ml, PTFE Beaker Cover, White
Price
$11.04
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
  • With pouring spout
  • Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.
Specifications
  • Capacity:
    1 ml
  • Color:
    White
  • Description:
    PTFE Beaker Cover