Orders LinkContactUsLinkComponent
Brushes, Red Sable, Electron Microscopy Sciences
Brushes, Red Sable, Electron Microscopy Sciences
Red Sable Brush Set of 3 Sizes (#2/0; #1 and #2), Sable hair, #2/0; #1 and #2
Catalog # 100496-236
Supplier #:  66105
Brushes, Red Sable, Electron Microscopy Sciences
Catalog # 100496-236
Brushes, Red Sable, Electron Microscopy Sciences
Brushes, Red Sable, Electron Microscopy Sciences
Red Sable Brush Set of 3 Sizes (#2/0; #1 and #2), Sable hair, #2/0; #1 and #2
Price
$42.70
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Specifications
  • Bristle material:
    Sable hair
  • Description:
    Red Sable Brush Set of 3 Sizes (#2/0; #1 and #2)
  • Size:
    #2/0; #1 and #2