MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with a location map for standard identification. Grains of standard materials are available and they measure approximately 2 x 2 mm in size. They are supplied with fully authenticated certificates of analysis.
Specifications
Specifications
- Description:Palladium Grain