Fine Mesh Grids, Electron Microscopy Sciences
Product Details & Documents
These fine mesh grids are carefully checked.
They are suitable for the low magnification range of a TEM.
Ordering information: Available as a sandwich in a folding 3.05 mm mesh grid, or in a square mesh 25 mm.
Specifications
- Type:2000
- Description:Mesh Grid
- Material:Copper